Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178078.1 | NP_001171549.1 | 3, 4 | 590 | ||
NM_001178079.1 | NP_001171550.1 | 3, 4 | 516 | |||
NM_001178080.1 | NP_001171551.1 | 2, 3 | 434 | |||
NM_001178081.1 | NP_001171552.1 | 1, 2 | 295 | |||
NM_003153.4 | NP_003144.3 | 3, 4 | 571 | |||
NR_033659.1 | 2, 3 | 432 | ||||
XM_006719575.2 | XP_006719638.1 | 4, 5 | 568 | |||
XM_011538703.2 | XP_011537005.1 | 3, 4 | 591 | |||
XM_011538704.2 | XP_011537006.1 | 4, 5 | 602 | |||
XM_011538705.2 | XP_011537007.1 | 4, 5 | 712 | |||
XM_011538706.2 | XP_011537008.1 | 4, 5 | 568 | |||
XM_011538707.2 | XP_011537009.1 | 3, 4 | 553 | |||
XM_011538708.2 | XP_011537010.1 | 2, 3 | 395 | |||
XM_011538709.2 | XP_011537011.1 | 3, 4 | 429 | |||
GenBank mRNA | AK290431.1 | - | 3, 4 | 519 | ||
AK291044.1 | - | 3, 4 | 517 | |||
AK298721.1 | - | 1, 2 | 115 | |||
AK299094.1 | - | 2, 3 | 368 | |||
AK308751.1 | - | 3, 4 | 414 | |||
AK312471.1 | - | 3, 4 | 437 | |||
AK316142.1 | - | 2, 3 | 378 | |||
BC075852.1 | - | 3, 4 | 372 | 5530399 | ||
BT007380.1 | - | 2, 3 | 265 | |||
U16031.1 | - | 3, 4 | 430 |