Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201334.1 | NP_001188263.1 | 6 | 833 | ||
NM_003203.4 | NP_003194.3 | 6 | 1065 | |||
XM_005264520.3 | XP_005264577.1 | 6 | 1090 | |||
XM_011533074.2 | XP_011531376.1 | 5 | 841 | |||
XM_011533075.1 | XP_011531377.1 | 6 | 1090 | |||
XM_017004787.1 | XP_016860276.1 | 6 | 1090 | |||
XM_017004788.1 | XP_016860277.1 | 6 | 1066 | |||
GenBank mRNA | AB026911.1 | - | 6 | 1003 | ||
AK097564.1 | - | 6 | 833 | |||
BC064559.1 | - | 6 | 962 | 5548897 | ||
EF158468.1 | - | 5 | 879 | |||
EF158469.1 | - | 3 | 403 |