Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003213.3 | NP_003204.2 | 13 | 1777 | ||
NM_201441.2 | NP_958849.1 | 12 | 1648 | |||
NM_201443.2 | NP_958851.1 | 11 | 1718 | |||
GenBank mRNA | AK222947.1 | - | 13 | 1664 | ||
BC015497.1 | - | 13 | 1675 | 3913870 | ||
BM555991.1 | - | 2 | 342 | |||
BX640921.1 | - | 11 | 1384 | |||
U63824.1 | - | 13 | 1649 |