Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145398.2 | NP_001138870.1 | 3 | 766 | ||
NM_003216.3 | NP_003207.1 | 3 | 772 | |||
GenBank mRNA | AK091916.1 | - | 3 | 644 | ||
AK295595.1 | - | 3 | 698 | |||
AK314906.1 | - | 3 | 688 | |||
BC039258.1 | - | 3 | 741 | 5286096 | ||
BC042476.2 | - | 3 | 742 | 4915095 | ||
BX537848.1 | - | 1 | 169 | |||
CR456592.1 | - | 3 | 669 | |||
CR541827.1 | - | 3 | 656 | |||
U06935.1 | - | 3 | 580 | |||
U44059.1 | - | 3 | 692 |