Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145398.2 | NP_001138870.1 | 4 | 3532 | ||
NM_003216.3 | NP_003207.1 | 4 | 3538 | |||
GenBank mRNA | AL122101.1 | - | 1 | 1751 | ||
BC039258.1 | - | 4 | 3505 | 5286096 | ||
BC042476.2 | - | 4 | 3508 | 4915095 | ||
BX537848.1 | - | 2 | 2935 |