Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003403.4 | NP_003394.1 | 1 | 914 | ||
GenBank mRNA | BC020324.1 | - | 1 | 509 | ||
BC037308.1 | - | 1 | 546 | 5261384 | ||
BC065366.1 | - | 1 | 479 | 5815774 | ||
M76541.1 | - | 1 | 508 | |||
M77698.1 | - | 1 | 674 | |||
Z14077.1 | - | 1 | 689 |