Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207024.1 | NP_001193953.1 | 5 | 1448 | ||
NM_002355.3 | NP_002346.1 | 7 | 1706 | |||
XM_005253376.2 | XP_005253433.1 | 7 | 1568 | |||
GenBank mRNA | BC017111.1 | - | 5 | 1106 | ||
BC024206.1 | - | 7 | 1598 | 3530887 | ||
BG750588.1 | - | 1 | 80 | |||
BM546062.1 | - | 2 | 861 | |||
M16985.1 | - | 7 | 1576 |