Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256523.1 | NP_001243452.1 | 4 | 2360 | ||
NM_001256524.1 | NP_001243453.1 | 6 | 2218 | |||
NM_001256525.1 | NP_001243454.1 | 5 | 2118 | |||
NM_003426.3 | NP_003417.2 | 5 | 2446 | |||
NR_046282.1 | 6 | 2542 | ||||
GenBank mRNA | AB209663.1 | - | 6 | 2254 | ||
AK128267.1 | - | 2 | 3451 | |||
AK291358.1 | - | 6 | 2144 | |||
AK299569.1 | - | 5 | 2044 | |||
BC013395.2 | - | 5 | 2044 | 3028001 | ||
BC048295.1 | - | 5 | 2031 | 5242841 | ||
CR456616.1 | - | 5 | 1985 | |||
X71623.1 | - | 4 | 1878 | |||
X92715.1 | - | 4 | 3044 |