Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003435.4 | NP_003426.3 | 3 | 1355 | ||
GenBank mRNA | AK291763.1 | - | 3 | 1324 | ||
BC011646.2 | - | 2 | 1214 | |||
BC021280.2 | - | 2 | 1199 | |||
BC042636.1 | - | 3 | 1314 | 4826076 | ||
BC112294.1 | - | 3 | 1115 | 8327762 | ||
BC113410.1 | - | 3 | 1112 | 8322462 | ||
U09412.1 | - | 3 | 1297 |
Gene Symbol: ZNF211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209847.1 | - | 1 | 707 |