Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172651.1 | NP_001166122.1 | 6 | 1386 | ||
NM_003451.2 | NP_003442.2 | 9 | 1201 | |||
GenBank mRNA | AK302274.1 | - | 5 | 1291 | ||
BC012012.1 | - | 5 | 792 | 4509206 | ||
U37263.1 | - | 9 | 1201 |
Gene Symbol: ZNF559-ZNF177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172650.2 | NP_001166121.1 | 12 | 1489 | ||
NM_001202425.1 | NP_001189354.1 | 9 | 1781 | |||
GenBank mRNA | U37251.1 | - | 9 | 1751 |