Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308007.1 | NP_001294936.1 | 3, 4 | 1207 | ||
NM_003797.4 | NP_003788.2 | 3, 4 | 1207 | |||
XM_005274373.2 | XP_005274430.1 | 3, 4 | 1047 | |||
XM_011545330.1 | XP_011543632.1 | 3, 4 | 1047 | |||
XM_011545331.1 | XP_011543633.1 | 3, 4 | 1047 | |||
XM_017018512.1 | XP_016874001.1 | 3, 4 | 1047 | |||
XM_017018513.1 | XP_016874002.1 | 3, 4 | 1047 | |||
GenBank mRNA | AF070418.1 | - | 3, 4 | 589 | ||
AF078933.1 | - | 3, 4 | 353 | |||
AF080227.1 | - | 3, 4 | 705 | |||
AF099032.1 | - | 3, 4 | 451 | |||
AK292120.1 | - | 3, 4 | 816 | |||
AK294133.1 | - | 3, 4 | 367 | |||
AK301265.1 | - | 3, 4 | 323 | |||
AL546955.3 | - | 3, 4 | 839 | |||
BC047672.2 | - | 3, 4 | 320 | |||
BC068995.1 | - | 3, 4 | 368 | 4637702 | ||
U90651.1 | - | 3, 4 | 451 |