Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261406.1 | NP_001248335.1 | 3 | 379 | ||
NM_001261407.1 | NP_001248336.1 | 3 | 379 | |||
NM_003829.4 | NP_003820.2 | 3 | 379 | |||
XM_005251622.4 | XP_005251679.1 | 3 | 328 | |||
XM_005251623.4 | XP_005251680.1 | 3 | 328 | |||
XM_006716885.3 | XP_006716948.1 | 3 | 377 | |||
XM_006716886.3 | XP_006716949.1 | 4 | 536 | |||
XM_006716887.3 | XP_006716950.1 | 3 | 480 | |||
XM_006716888.3 | XP_006716951.1 | 3 | 328 | |||
XM_006716889.3 | XP_006716952.1 | 3 | 331 | |||
XM_006716891.3 | XP_006716954.1 | 3 | 331 | |||
XM_017015252.1 | XP_016870741.1 | 3 | 328 | |||
XM_017015253.1 | XP_016870742.1 | 3 | 316 | |||
XM_017015254.1 | XP_016870743.1 | 4 | 539 | |||
XM_017015255.1 | XP_016870744.1 | 3 | 387 | |||
XM_017015256.1 | XP_016870745.1 | 3 | 328 | |||
XM_017015257.1 | XP_016870746.1 | 3 | 315 | |||
XM_017015258.1 | XP_016870747.1 | 3 | 328 | |||
XM_017015259.1 | XP_016870748.1 | 3 | 328 | |||
GenBank mRNA | AB210041.1 | - | 3 | 380 | ||
AF093419.1 | - | 2 | 203 | |||
BC140793.1 | - | 2 | 203 | 9021661 | ||
BC144564.1 | - | 2 | 203 | 9053092 |