Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281453.1 | NP_001268382.1 | 7 | 1651 | ||
NM_001281454.1 | NP_001268383.1 | 7 | 1555 | |||
GenBank mRNA | AF072247.1 | - | 7 | 1545 | ||
AK001474.1 | - | 6 | 1351 | |||
AK021532.1 | - | 6 | 1302 | |||
BC000872.1 | - | 1 | 119 | |||
BC009372.1 | - | 7 | 1487 | 4122128 | ||
BC009438.1 | - | 4 | 1346 | |||
BC018784.1 | - | 1 | 654 | |||
BC032443.1 | - | 5 | 1159 | |||
BC043619.1 | - | 7 | 1505 | 6156267 |