Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004229.3 | NP_004220.2 | 1 | 343 | ||
XM_017029962.1 | XP_016885451.1 | 1 | 115 | |||
GenBank mRNA | AB006651.1 | - | 1 | 322 | ||
AF104256.1 | - | 1 | 203 | |||
AF135802.1 | - | 1 | 276 | |||
AF304448.1 | - | 1 | 322 | |||
AK293070.1 | - | 1 | 521 | |||
BC098377.1 | - | 1 | 343 | 30527948 | ||
BC132672.1 | - | 1 | 312 | 40146694 | ||
BC132674.1 | - | 1 | 312 | 40146696 | ||
HG518165.2 | - | 1 | 45 |