Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314052.1 | NP_001300981.1 | 4 | 2470 | ||
NM_004235.5 | NP_004226.3 | 5 | 2368 | |||
GenBank mRNA | AF105036.1 | - | 5 | 2122 | ||
AK095134.1 | - | 3 | 1874 | |||
AK095894.1 | - | 1 | 1617 | |||
BC029923.1 | - | 5 | 1933 | 5111134 | ||
BC030811.1 | - | 3 | 1924 | 4703002 |