Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004561.3 | NP_004552.2 | 4 | 948 | ||
XM_005274018.3 | XP_005274075.1 | 4 | 766 | |||
XM_011545067.2 | XP_011543369.1 | 4 | 764 | |||
XM_011545068.2 | XP_011543370.1 | 4 | 1309 | |||
XM_017017837.1 | XP_016873326.1 | 4 | 1602 | |||
XM_017017838.1 | XP_016873327.1 | 4 | 641 | |||
GenBank mRNA | AF016045.1 | - | 3 | 828 | ||
AK291711.1 | - | 4 | 883 | |||
BC043146.1 | - | 4 | 539 | |||
BC059408.1 | - | 4 | 924 | 30343789 | ||
BX648725.1 | - | 4 | 828 |