Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281432.1 | NP_001268361.1 | 3 | 247 | ||
NM_001281433.1 | NP_001268362.1 | 3 | 247 | |||
NM_004773.3 | NP_004764.1 | 3 | 247 | |||
NR_104009.1 | 3 | 242 | ||||
NR_104010.1 | 2 | 215 | ||||
NR_104011.1 | 2 | 215 | ||||
GenBank mRNA | AB209193.1 | - | 1 | 3711 | ||
AF400652.1 | - | 3 | 239 | |||
AK290858.1 | - | 3 | 198 | |||
AK308583.1 | - | 2 | 143 | |||
AK310022.1 | - | 3 | 182 | |||
AV763423.1 | - | 214 | ||||
BC017931.1 | - | 3 | 212 | 4669658 | ||
BC105608.1 | - | 2 | 187 | |||
BG032116.1 | - | 3 | 182 | |||
BI767446.1 | - | 3 | 180 | |||
BM457955.1 | - | 177 | ||||
CB996852.1 | - | 3 | 272 | |||
HY045632.1 | - | 3 | 248 | |||
L40410.1 | - | 3 | 165 |