Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005121.2 | NP_005112.2 | 3 | 449 | ||
XM_011525551.2 | XP_011523853.1 | 3 | 1150 | |||
XM_011525552.2 | XP_011523854.1 | 3 | 1150 | |||
GenBank mRNA | AF117754.1 | - | 4 | 449 | ||
BC140891.1 | - | 3 | 423 | 9021759 | ||
BP332524.1 | - | 3 | 449 | |||
BQ709424.1 | - | 2 | 205 |