Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005171.4 | NP_005162.1 | 5, 6 | 814 | ||
XM_011538386.2 | XP_011536688.1 | 5, 6 | 2871 | |||
XM_011538387.2 | XP_011536689.1 | 5, 6 | 544 | |||
XM_011538388.2 | XP_011536690.1 | 5, 6 | 745 | |||
XM_017019331.1 | XP_016874820.1 | 5, 6 | 584 | |||
XM_017019332.1 | XP_016874821.1 | 5, 6 | 678 | |||
XM_017019333.1 | XP_016874822.1 | 5, 6 | 564 | |||
XM_017019334.1 | XP_016874823.1 | 5, 6 | 590 | |||
XM_017019335.1 | XP_016874824.1 | 5, 6 | 588 | |||
XM_017019336.1 | XP_016874825.1 | 5, 6 | 583 | |||
GenBank mRNA | AK299240.1 | - | 4, 5 | 430 | ||
AK313406.1 | - | 5, 6 | 719 | |||
BC029619.1 | - | 5, 6 | 528 | 4102231 | ||
BG502726.1 | - | 5, 6 | 547 | |||
BG611151.1 | - | 1, 2 | 30 | |||
BX102171.1 | - | 1, 2 | 50 |