Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278647.1 | NP_001265576.1 | 7 | 1415 | ||
NM_001278648.1 | NP_001265577.1 | 7 | 1436 | |||
NM_005341.3 | NP_005332.1 | 7 | 1502 | |||
XM_017001112.1 | XP_016856601.1 | 7 | 1592 | |||
XM_017001113.1 | XP_016856602.1 | 7 | 1533 | |||
XM_017001114.1 | XP_016856603.1 | 6 | 1308 | |||
GenBank mRNA | AF318381.1 | - | 2 | 1148 | ||
AK290156.1 | - | 7 | 1410 | |||
AK312786.1 | - | 7 | 1418 | |||
BC013573.2 | - | 6 | 1361 | 3906038 | ||
BX648395.1 | - | 3 | 1637 | |||
L16896.1 | - | 7 | 1442 |