Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301228.1 | NP_001288157.1 | 5 | 1016 | ||
NM_001301229.1 | NP_001288158.1 | 5 | 907 | |||
NM_001301231.1 | NP_001288160.1 | 5 | 946 | |||
NM_005342.3 | NP_005333.2 | 5 | 911 | |||
GenBank mRNA | BX537505.1 | - | 3 | 2307 | ||
Y10043.1 | - | 5 | 890 |