Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYB SIRNA Assays ›
Gene Symbol:
MYB
Gene Name:
MYB proto-oncogene, transcription factor
Gene Aliases:
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location:
Chr.6: 135180981 - 135219173 on Build GRCh38
UniGene ID:
Hs.606320
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115652
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130172.1
NP_001123644.1
2
276
NM_001130173.1
NP_001123645.1
2
276
NM_001161656.1
NP_001155128.1
2
276
NM_001161657.1
NP_001155129.1
2
276
NM_001161658.1
NP_001155130.1
2
276
NM_001161659.1
NP_001155131.1
2
276
NM_001161660.1
NP_001155132.1
2
276
NM_005375.3
NP_005366.2
2
276
NR_134958.1
2
276
NR_134959.1
2
276
NR_134960.1
2
276
NR_134961.1
2
276
NR_134962.1
2
276
NR_134963.1
2
276
NR_134964.1
2
276
NR_134965.1
2
276
GenBank mRNA
AF104863.1
-
2
154
AJ606317.1
-
2
276
AJ606318.1
-
2
276
AJ606319.1
-
2
276
AJ606320.1
-
2
276
AJ606321.1
-
2
276
AJ606322.1
-
2
276
AJ606323.1
-
2
276
AJ606324.1
-
2
276
AJ616235.1
-
2
191
AJ616792.1
-
1
54
AY787443.1
-
2
77
AY787444.1
-
2
77
AY787445.1
-
2
77
AY787446.1
-
2
77
AY787447.1
-
2
77
AY787448.1
-
2
77
AY787449.1
-
2
77
AY787450.1
-
2
77
AY787451.1
-
2
77
AY787452.1
-
2
77
AY787453.1
-
2
77
AY787454.1
-
2
77
AY787455.1
-
2
77
AY787456.1
-
2
77
AY787457.1
-
2
77
AY787458.1
-
2
77
AY787459.1
-
2
77
AY787460.1
-
2
77
AY787461.1
-
2
77
AY787462.1
-
2
77
AY787463.1
-
2
77
AY787464.1
-
2
77
AY787465.1
-
2
77
AY787466.1
-
2
77
AY787467.1
-
2
77
AY787468.1
-
2
77
AY787469.1
-
2
77
AY787470.1
-
2
77
AY787471.1
-
2
77
AY787472.1
-
2
77
AY787473.1
-
2
77
AY787474.1
-
2
77
AY787475.1
-
2
77
BC064955.1
-
2
228
6069320
M15024.1
-
2
190
X52125.1
-
2
274
X52126.1
-
1
80
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
chromatin remodeling
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of T-helper cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of histone H3-K4 methylation
positive regulation of histone H3-K9 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline