Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134673.3 | NP_001128145.1 | 2 | 745 | ||
NM_001145511.1 | NP_001138983.1 | 2 | 589 | |||
NM_001145512.1 | NP_001138984.1 | 3 | 478 | |||
NM_005595.4 | NP_005586.1 | 2 | 745 | |||
XM_011541512.2 | XP_011539814.1 | 2 | 806 | |||
XM_011541514.2 | XP_011539816.1 | 2 | 739 | |||
XM_011541515.2 | XP_011539817.1 | 2 | 806 | |||
XM_017001362.1 | XP_016856851.1 | 2 | 667 | |||
GenBank mRNA | AB037860.1 | - | 2 | 745 | ||
AK299289.1 | - | 2 | 589 | |||
AK299356.1 | - | 2 | 486 | |||
AK299579.1 | - | 3 | 478 | |||
BC022264.1 | - | 2 | 432 | 4270934 | ||
U07809.1 | - | 1 | 208 |