Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324339.1 | NP_001311268.1 | 4 | 494 | ||
NM_005649.2 | NP_005640.2 | 5 | 578 | |||
XM_011534645.2 | XP_011532947.1 | 5 | 1085 | |||
XM_017009791.1 | XP_016865280.1 | 5 | 483 | |||
GenBank mRNA | AF116030.1 | - | 5 | 584 | ||
AK290711.1 | - | 5 | 473 | |||
BC047105.1 | - | 5 | 535 | 5312219 | ||
D89928.1 | - | 5 | 532 | |||
EU794664.1 | - | 5 | 578 |