Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002412.4 | NP_002403.2 | 4 | 445 | ||
XM_005252682.2 | XP_005252739.1 | 4 | 482 | |||
XM_017016275.1 | XP_016871764.1 | 3 | 268 | |||
GenBank mRNA | AK293593.1 | - | 4 | 415 | ||
BC000824.1 | - | 4 | 373 | 3454207 | ||
BI520029.1 | - | 4 | 427 | |||
BT006714.1 | - | 3 | 320 | |||
DC387686.1 | - | 4 | 439 | |||
DC416206.1 | - | 4 | 445 | |||
KC249995.1 | - | 3 | 354 | |||
M29971.1 | - | 4 | 416 | |||
M31767.1 | - | 3 | 342 | |||
M60761.1 | - | 3 | 335 | |||
X54228.1 | - | 3 | 360 |