Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002416.2 | NP_002407.1 | 4 | 2383 | ||
GenBank mRNA | BC025745.1 | - | 4 | 2358 | ||
BC042178.1 | - | 4 | 2331 | |||
BC063122.1 | - | 4 | 2374 | 30370826 | ||
BC095396.1 | - | 4 | 2369 | 30336410 | ||
X72755.1 | - | 4 | 2348 |