Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145102.1 | NP_001138574.1 | 5, 6 | 714 | ||
NM_001145103.1 | NP_001138575.1 | 5, 6 | 525 | |||
NM_001145104.1 | NP_001138576.1 | 3, 4 | 158 | |||
NM_005902.3 | NP_005893.1 | 5, 6 | 949 | |||
XM_011521559.2 | XP_011519861.1 | 4, 5 | 976 | |||
XM_011521560.1 | XP_011519862.1 | 5, 6 | 888 | |||
GenBank mRNA | AB209616.1 | - | 5, 6 | 534 | ||
AK290881.1 | - | 6, 7 | 874 | |||
AK298139.1 | - | 5, 6 | 525 | |||
AK300614.1 | - | 3, 4 | 155 | |||
AK301348.1 | - | 3, 4 | 150 | |||
AK316017.1 | - | 5, 6 | 574 | |||
BC050743.1 | - | 5, 6 | 867 | 6094328 | ||
DA977882.1 | - | 3, 4 | 158 | |||
U68019.1 | - | 5, 6 | 717 | |||
U76622.1 | - | 5, 6 | 688 |