Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145966.1 | NP_001139438.1 | 10, 11 | 1370 | ||
NM_002417.4 | NP_002408.3 | 11, 12 | 2450 | |||
XM_011539818.2 | XP_011538120.1 | 8, 9 | 2181 | |||
GenBank mRNA | AJ567756.1 | - | 7, 8 | 980 | ||
X65550.1 | - | 11, 12 | 2451 | |||
X65551.1 | - | 10, 11 | 1371 |