Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001419.2 | NP_001001419.1 | 9 | 2261 | ||
NM_001001420.2 | NP_001001420.1 | 7 | 2102 | |||
NM_005903.6 | NP_005894.3 | 8 | 2177 | |||
XM_017009470.1 | XP_016864959.1 | 9 | 2393 | |||
GenBank mRNA | AF010601.1 | - | 6 | 1946 | ||
AK311231.1 | - | 4 | 624 | |||
BC009682.2 | - | 6 | 1983 | 3906006 | ||
BX648328.1 | - | 8 | 2116 | |||
CR749366.1 | - | 8 | 2116 | |||
CR749473.1 | - | 8 | 2104 | |||
U59913.1 | - | 6 | 1942 |