Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170931.1 | NP_001164402.1 | 3 | 971 | ||
NM_005938.3 | NP_005929.2 | 2 | 1136 | |||
XM_011530957.2 | XP_011529259.1 | 2 | 553 | |||
GenBank mRNA | AF384029.1 | - | 3 | 591 | ||
AK300142.1 | - | 2 | 1082 | |||
BC026735.1 | - | 1 | 245 | |||
BC106761.2 | - | 2 | 862 | 40025381 | ||
X93996.1 | - | 2 | 926 |