Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005967.3 | NP_005958.1 | 7 | 2338 | ||
XM_005268894.2 | XP_005268951.1 | 6 | 2146 | |||
GenBank mRNA | BC007756.2 | - | 6 | 1498 | ||
BC065931.1 | - | 7 | 2125 | 6157017 | ||
X70991.1 | - | 6 | 1812 |