Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145938.1 | NP_001139410.1 | 8, 9 | 1195 | ||
NM_002421.3 | NP_002412.1 | 8, 9 | 1347 | |||
GenBank mRNA | AK024818.1 | - | 8, 9 | 1270 | ||
AK098450.1 | - | 9, 10 | 1202 | |||
AK130687.1 | - | 8, 9 | 1270 | |||
AK223034.1 | - | 7, 8 | 973 | |||
AK223035.1 | - | 8, 9 | 1228 | |||
AK223036.1 | - | 6, 7 | 811 | |||
AK223056.1 | - | 7, 8 | 1013 | |||
AK223281.1 | - | 8, 9 | 1270 | |||
AK225495.1 | - | 7, 8 | 1038 | |||
AK297723.1 | - | 8, 9 | 1118 | |||
BC013118.1 | - | 4, 5 | 495 | |||
BC013875.2 | - | 8, 9 | 1246 | 3834572 | ||
BT006874.1 | - | 8, 9 | 1204 | |||
BT020147.1 | - | 8, 9 | 1204 | |||
M13509.1 | - | 8, 9 | 1272 | |||
X05231.1 | - | 8, 9 | 1270 | |||
X54925.1 | - | 8, 9 | 1272 |