Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143835.1 | NP_001137307.1 | 5 | 653 | ||
NM_006165.3 | NP_006156.2 | 3 | 612 | |||
XM_005271567.3 | XP_005271624.1 | 5 | 653 | |||
XM_005271568.3 | XP_005271625.1 | 4 | 568 | |||
XM_011542850.2 | XP_011541152.1 | 6 | 690 | |||
XM_011542851.2 | XP_011541153.1 | 5 | 672 | |||
XM_011542852.2 | XP_011541154.1 | 4 | 510 | |||
XM_017017793.1 | XP_016873282.1 | 4 | 564 | |||
XM_017017794.1 | XP_016873283.1 | 4 | 564 | |||
XM_017017795.1 | XP_016873284.1 | 6 | 690 | |||
XM_017017796.1 | XP_016873285.1 | 4 | 568 | |||
XM_017017797.1 | XP_016873286.1 | 4 | 564 | |||
XM_017017798.1 | XP_016873287.1 | 5 | 653 | |||
XM_017017799.1 | XP_016873288.1 | 5 | 654 | |||
XM_017017800.1 | XP_016873289.1 | 5 | 630 | |||
GenBank mRNA | BC063280.1 | - | 5 | 599 | 5271534 | |
X80878.1 | - | 3 | 621 |