Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006166.3 | NP_006157.1 | 8 | 2684 | ||
XM_005268908.4 | XP_005268965.1 | 9 | 2748 | |||
XM_005268909.4 | XP_005268966.1 | 9 | 2731 | |||
XM_005268910.4 | XP_005268967.1 | 9 | 3403 |