Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077261.3 | NP_001070729.2 | 8 | 1047 | ||
NM_001206654.1 | NP_001193583.1 | 8 | 1047 | |||
NM_006312.5 | NP_006303.4 | 8 | 1047 | |||
GenBank mRNA | AB209089.1 | - | 8 | 1058 | ||
AF113003.1 | - | 6 | 727 | |||
AF125672.1 | - | 7 | 882 | |||
AY965853.1 | - | 7 | 882 |