Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077261.3 | NP_001070729.2 | 48 | 8661 | ||
NM_001206654.1 | NP_001193583.1 | 48 | 8799 | |||
NM_006312.5 | NP_006303.4 | 49 | 8829 | |||
GenBank mRNA | AF113003.1 | - | 48 | 8519 | ||
AF125672.1 | - | 47 | 8644 | |||
AY965853.1 | - | 47 | 8506 | |||
BC004326.1 | - | 10 | 2785 | |||
BM687540.1 | - | 1 | 341 | |||
L32841.1 | - | 1 | 72 | |||
X79269.1 | - | 1 | 72 |