Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLAGL1 SIRNA Assays ›
Gene Symbol:
PLAGL1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
LOT1, ZAC, ZAC1
Chromosome Location:
Chr.6: 143940300 - 144064599 on Build GRCh38
UniGene ID:
Hs.743225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115818
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLAGL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080951.2
NP_001074420.1
8
2374
NM_001080952.2
NP_001074421.1
7
2526
NM_001080953.2
NP_001074422.1
7
2480
NM_001080954.2
NP_001074423.1
7
2479
NM_001080955.2
NP_001074424.1
6
1823
NM_001080956.2
NP_001074425.1
6
2018
NM_001289037.1
NP_001275966.1
5
1782
NM_001289038.1
NP_001275967.1
4
1736
NM_001289039.1
NP_001275968.1
5
1839
NM_001289040.1
NP_001275969.1
6
1808
NM_001289041.1
NP_001275970.1
6
2018
NM_001289042.1
NP_001275971.1
5
2170
NM_001289043.1
NP_001275972.1
7
2293
NM_001289044.1
NP_001275973.1
6
2244
NM_001289045.1
NP_001275974.1
7
2788
NM_001289046.1
NP_001275975.1
9
2769
NM_001289047.1
NP_001275976.1
4
2333
NM_001289048.1
NP_001275977.1
7
2494
NM_001289049.1
NP_001275978.1
6
2439
NM_001317156.1
NP_001304085.1
4
2700
NM_001317157.1
NP_001304086.1
3
2876
NM_001317158.1
NP_001304087.1
3
2238
NM_001317159.1
NP_001304088.1
8
2360
NM_001317160.1
NP_001304089.1
6
2004
NM_001317161.1
NP_001304090.1
7
2494
NM_001317162.1
NP_001304091.1
8
2520
NM_006718.4
NP_006709.2
7
2512
GenBank mRNA
AJ303119.1
-
2
1546
AJ311395.1
-
9
2759
AK091707.1
-
4
2700
AK096331.1
-
7
2755
BC109085.2
-
3
1966
40034491
BC109086.2
-
3
1966
40034497
BX537397.1
-
3
5207
CR749329.1
-
3
5391
U72621.4
-
6
3851
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
cell differentiation
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline