Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002432.1 | NP_002423.1 | 5 | 1000 | ||
GenBank mRNA | AK290392.1 | - | 5 | 1007 | ||
AK313210.1 | - | 5 | 1011 | |||
BC020715.1 | - | 5 | 1038 | |||
BC032319.1 | - | 5 | 1011 | 5223430 | ||
M81750.1 | - | 5 | 1000 |