Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193646.1 | NP_001180575.1 | 3 | 592 | ||
NM_001290746.1 | NP_001277675.1 | 3 | 563 | |||
NM_012068.5 | NP_036200.2 | 4 | 797 | |||
XM_011526629.2 | XP_011524931.1 | 3 | 499 | |||
GenBank mRNA | AB021663.2 | - | 3 | 534 | ||
AB073613.1 | - | 3 | 530 | |||
AF305687.1 | - | 2 | 265 | |||
AK024402.1 | - | 3 | 534 | |||
AK290592.1 | - | 4 | 736 | |||
BC005174.2 | - | 3 | 521 | 3504701 | ||
BF528888.1 | - | 3 | 588 | |||
BM462249.1 | - | 3 | 433 | |||
BU187153.1 | - | 3 | 517 |