Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002442.3 | NP_002433.1 | 6 | 466 | ||
XM_006719403.3 | XP_006719466.1 | 6 | 460 | |||
XM_011538361.2 | XP_011536663.1 | 6 | 460 | |||
XM_011538362.2 | XP_011536664.1 | 6 | 460 | |||
XM_011538363.2 | XP_011536665.1 | 6 | 460 | |||
XM_011538364.2 | XP_011536666.1 | 2 | 1010 | |||
XM_011538365.2 | XP_011536667.1 | 3 | 129 | |||
XM_011538366.2 | XP_011536668.1 | 3 | 1137 | |||
GenBank mRNA | AB012851.1 | - | 6 | 441 | ||
AB072590.1 | - | 3 | 188 | |||
AB072591.1 | - | 3 | 188 | |||
AB072592.1 | - | 3 | 188 | |||
AB073212.1 | - | 3 | 181 | |||
BX393884.2 | - | 6 | 466 |