Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002444.2 | NP_002435.1 | 2, 3 | 302 | ||
XM_005262269.2 | XP_005262326.1 | 2, 3 | 323 | |||
XM_011530959.1 | XP_011529261.1 | 2, 3 | 218 | |||
XM_017029545.1 | XP_016885034.1 | 4, 5 | 446 | |||
XM_017029546.1 | XP_016885035.1 | 3, 4 | 342 | |||
GenBank mRNA | AK308486.1 | - | 2, 3 | 265 | ||
AK309354.1 | - | 1, 2 | 67 | |||
BC001112.2 | - | 2, 3 | 274 | |||
BC011827.2 | - | 2, 3 | 274 | |||
BC017293.2 | - | 2, 3 | 255 | 4908580 | ||
CN386296.1 | - | 2, 3 | 302 | |||
EU794636.1 | - | 2, 3 | 255 | |||
M69066.1 | - | 2, 3 | 204 |