Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012384.4 | NP_036516.1 | 3 | 421 | ||
XM_005260202.4 | XP_005260259.1 | 3 | 632 | |||
XM_006723776.2 | XP_006723839.1 | 1 | 78 | |||
XM_011528779.2 | XP_011527081.1 | 1 | 68 | |||
GenBank mRNA | AB033095.1 | - | 3 | 298 | ||
AF173867.1 | - | 2 | 697 | |||
AK298927.1 | - | 3 | 418 | |||
AL133646.1 | - | 1 | 150 | |||
BC036305.1 | - | 3 | 348 | 5166938 | ||
BQ954281.1 | - | 2 | 123 | |||
DB474085.1 | - | 3 | 374 |