Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012384.4 | NP_036516.1 | 10 | 3769 | ||
XM_005260202.4 | XP_005260259.1 | 10 | 3980 | |||
XM_006723776.2 | XP_006723839.1 | 8 | 3421 | |||
XM_011528779.2 | XP_011527081.1 | 8 | 3485 | |||
GenBank mRNA | AB033095.1 | - | 10 | 3646 | ||
AL117510.1 | - | 1 | 2149 | |||
AL133646.1 | - | 8 | 3493 |