Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278508.2 | NP_001265437.2 | 4 | 616 | ||
NM_001278509.2 | NP_001265438.2 | 5 | 619 | |||
NM_001288759.2 | NP_001275688.2 | 5 | 696 | |||
NM_001288760.2 | NP_001275689.1 | 7 | 864 | |||
NM_001288761.2 | NP_001275690.1 | 4 | 492 | |||
NM_001288762.2 | NP_001275691.1 | 4 | 569 | |||
NM_001291633.1 | NP_001278562.1 | 5 | 614 | |||
NM_013256.5 | NP_037388.3 | 5 | 691 | |||
XM_011527280.2 | XP_011525582.1 | 5 | 3024 | |||
XM_017027250.1 | XP_016882739.1 | 5 | 765 | |||
XM_017027251.1 | XP_016882740.1 | 6 | 834 | |||
XM_017027252.1 | XP_016882741.1 | 6 | 740 | |||
XM_017027253.1 | XP_016882742.1 | 6 | 809 | |||
XM_017027254.1 | XP_016882743.1 | 4 | 375 | |||
XM_017027255.1 | XP_016882744.1 | 6 | 827 | |||
XM_017027256.1 | XP_016882745.1 | 6 | 879 | |||
GenBank mRNA | AF192913.1 | - | 6 | 673 | ||
AK024047.1 | - | 5 | 616 | |||
AK122688.1 | - | 4 | 613 | |||
AK315193.1 | - | 5 | 690 | |||
AU132370.1 | - | 5 | 611 | |||
BC033642.1 | - | 5 | 683 | 5527054 | ||
BC113015.1 | - | 5 | 604 | 40034649 | ||
BP238630.1 | - | 4 | 489 |