Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013448.2 | NP_038476.2 | 3 | 764 | ||
NM_182648.1 | NP_872589.1 | 3 | 764 | |||
GenBank mRNA | AB032252.1 | - | 3 | 761 | ||
AF086397.1 | - | 3 | 362 | |||
AF161435.1 | - | 3 | 450 | |||
AF213467.1 | - | 3 | 362 | |||
AF221130.1 | - | 2 | 196 | |||
BC020636.1 | - | 2 | 524 | |||
BC108711.1 | - | 3 | 428 |