Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014159.6 | NP_054878.5 | 3 | 1934 | ||
XM_011533631.2 | XP_011531933.1 | 4 | 3430 | |||
XM_011533632.2 | XP_011531934.1 | 3 | 2006 | |||
XM_011533633.2 | XP_011531935.1 | 4 | 3432 | |||
XM_011533634.1 | XP_011531936.1 | 3 | 1959 | |||
XM_017006270.1 | XP_016861759.1 | 4 | 3433 | |||
GenBank mRNA | AJ238403.1 | - | 1 | 781 | ||
AK091413.1 | - | 1 | 598 | |||
AK127782.1 | - | 1 | 1351 | |||
AK128096.1 | - | 3 | 1959 | |||
AK131371.1 | - | 3 | 1924 | |||
AL831959.1 | - | 1 | 1033 | |||
AY576987.1 | - | 1 | 443 | |||
AY576988.1 | - | 1 | 443 | |||
BC072440.1 | - | 1 | 722 | |||
BC117162.1 | - | 1 | 557 | 40125713 | ||
BC117164.1 | - | 1 | 557 | 40125715 | ||
BX649110.1 | - | 1 | 1496 |