Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126335.1 | NP_001119807.1 | 12, 13 | 1526 | ||
NM_001243036.1 | NP_001229965.1 | 12, 13 | 1621 | |||
NM_014270.4 | NP_055085.1 | 12, 13 | 1612 | |||
XM_006722992.1 | XP_006723055.1 | 10, 11 | 1287 | |||
XM_011526402.2 | XP_011524704.1 | 12, 13 | 2716 | |||
XM_017026230.1 | XP_016881719.1 | 10, 11 | 1262 | |||
GenBank mRNA | AB033548.1 | - | 12, 13 | 1497 | ||
AF141289.1 | - | 12, 13 | 1594 | |||
AJ249199.1 | - | 11, 12 | 1471 | |||
AK026446.1 | - | 12, 13 | 1595 | |||
AK223147.1 | - | 12, 13 | 1600 | |||
AK313708.1 | - | 12, 13 | 1513 | |||
AL365340.1 | - | 5, 6 | 664 | |||
AL365341.1 | - | 4, 5 | 448 | |||
AY170373.1 | - | 7, 8 | 943 | |||
BC017962.1 | - | 12, 13 | 1601 | 4608486 |