Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFCP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014553.2 | NP_055368.1 | 7 | 768 | ||
XM_017003902.1 | XP_016859391.1 | 7 | 776 | |||
XM_017003904.1 | XP_016859393.1 | 7 | 776 | |||
XM_017003905.1 | XP_016859394.1 | 7 | 776 | |||
GenBank mRNA | AF198488.1 | - | 7 | 765 | ||
AL137740.1 | - | 7 | 768 | |||
BC064698.1 | - | 7 | 768 | 30347863 |