Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014564.4 | NP_055379.1 | 6 | 1614 | ||
NM_178138.5 | NP_835258.1 | 6 | 1582 | |||
XM_005263410.1 | XP_005263467.1 | 6 | 1896 | |||
XM_017015168.1 | XP_016870657.1 | 5 | 1393 | |||
GenBank mRNA | AB593062.1 | - | 6 | 1582 | ||
AB593063.1 | - | 6 | 1581 | |||
AB593064.1 | - | 6 | 1896 | |||
AB593065.1 | - | 5 | 2289 | |||
AB593066.1 | - | 6 | 1614 | |||
AB593067.1 | - | 6 | 1579 | |||
AF156888.1 | - | 6 | 1569 | |||
AF156889.1 | - | 6 | 1599 |