Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002471.3 | NP_002462.2 | 3 | 208 | ||
GenBank mRNA | AK309494.1 | - | 3 | 204 | ||
BC132667.1 | - | 2 | 167 | 40146689 | ||
BX501231.1 | - | 3 | 207 | |||
D00943.1 | - | 1 | 137 | |||
DA566231.1 | - | 3 | 206 |